Learn More
EMD Millipore™ Novagen™ KOD Hot Start DNA Polymerase Available on GSA/VA Contract for Federal Government customers only.
For PCR amplification of long strand and GC-rich DNA templates, cloning and cDNA amplification applications
Manufacturer:  EMD Millipore™ 710863
 View more versions of this product
Catalog No. 71-086-3
  Check Availability
Add to cart
Disclaimers:
For Research Use Only
Specifications
Description
  • High accuracy, yield, and processivity of commercially available proofreading DNA polymerases
  • Amplifies genomic DNA templates up to 12 kbp
  • Amplifies plasmid and lambda DNA templates up to 21 kbp
  • Successfully amplifies GC-rich sequences
  • Eliminates mispriming and primer-dimer formation
  • Convenient room-temperature setup compatible with automation
  • Optimal KOD Hot Start Buffer for PCR performance over wide range of targets
Description & Specifications
Specifications
KOD Hot Start DNA Polymerase
Recombinant Thermococcus kodakaraensis KOD1 DNA polymerase expressed in E. coli
-20°C
1U/μL
200U