Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Novagen™ LB Agar, Miller

Used as granulated medium for cultivation of E. coli. MilliporeSigma™; Novagen™ LB Agar, Miller granules ensure rapid and uniform dissolution in water.

Manufacturer:  MilliporeSigma™ 717525

 View more versions of this product

Catalog No. 71-752-5


Add to cart

Description

Description

  • Prevents clumping of medium and inhalation of airborne powder
  • Supplied in several formats
  • EasyPak is aluminum foil pouch containing 37g granulated medium sufficient for 1L culture
  • Add EasyPak contents to 1L water, microwave to dissolve agar and sterilize
  • Supplied in 500g and 5kg bottles
  • Composition of LB Agar MILLER per liter is 5g yeast extract, 10g peptone from casein, 10g sodium chloride, and 12g agar-agar
Specifications

Specifications

LB Agar
500 g
LB Agar
Safety and Handling

Safety and Handling

SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit