Learn More
MilliporeSigma™ Novagen™ NanoJuice™, Transfection Kit
Designed for efficient transfection primary and other mammalian cells, uses cutting-edge nanotechnology of Priostar™ dendrimers.
$606.50
Specifications
Format Liquid
View More Specs
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
71-902-4 View Documents MilliporeSigma™
719024
2400 Reactions/Pk. Each for $606.50
Add to cart
71-902-3 View Documents MilliporeSigma™
719023
240 Reactions/Pk. This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description
Description
  • Suitable for stable and transient transfections
  • Minimal cellular toxicity
  • Polycationic liposomal formulation
  • Optimize transfection efficiencies and reduces cytotoxicity
  • Compatible with both serum-containing and serum-free media
Specifications
Specifications
Liquid