Learn More
MilliporeSigma™ Novagen™ NovaTaq™ Hot Start Master Mix Kits
Premixed 2X hot start PCR components for convenience and reproducibility
Supplier: MilliporeSigma™ 716764
Description
- Simplifies set-up for PCR and offers time savings, consistency, and minimal risk of contamination
- Provide sufficient components for 200 or 1000 standard 50μL amplification reactions
Specifications
Ready-to-use 2X mixture of NovaTaq Hot Start DNA Polymerase, ultrapure deoxynucleotides, and reaction buffer with MgCl2, PCR Grade Water, MgCl2 for optimizing Mg2+ concentrations | |
Hot start master mix kit | |
1000 rxn |