Learn More
MilliporeSigma™ Novagen™ Overnight Express™ Autoinduction System 2
Used for expression of proteins in multiple cultures.
Supplier:  MilliporeSigma™ 713663
 View more versions of this product
Catalog No. 71-366-3
Add to Cart
Includes:
  • 20mL OnEx™ Solution 1
  • 50mL OnEx Solution 2
  • 1mL OnEx Solution 3
  • 1mL OnEx Solution 4
  • 20mL OnEx Solution 5
  • 20mL OnEx Solution 6
Description
Description
EMD Millipore Novagen™ Overnight Express™ Autoinduction System 2 provides complete, chemically defined medium for high-level protein expression with pET System.
  • High cell densities and protein expression levels
  • Chemically defined medium, minimal variability
  • No need for monitor cell growth rate or add inducer
  • Used for Se-Met labeling of proteins to be crystallized for x-ray diffraction studies
  • Induction of numerous expression clones simultaneously
  • Compatible with cultures grown in flasks, culture tubes, and deep-well plates
  • Minimal sample handling
  • Packaging: Glass Bottle
Specifications
Specifications
Multiple storage requirements
1 L Kit
Product Suggestions
Product Suggestions