Learn More
MilliporeSigma™ Novagen™ Strep·Tactin™ Superflow™ Agarose
Used for gravity flow, low pressure and FPLC chromatography.
Manufacturer:  MilliporeSigma™ 715923
 View more versions of this product
Catalog No. 71-592-3
Add to cart
Description
Description
  • Cross-linked agarose derivatized with StrepTactin protein
  • Binding capacity: 50 to 100nmol/mL settled resin (3mg of 30 kd protein per mL settled resin)
  • Applications:
    • Optimized for column affinity chromatography
    • Packaging: Glass Bottle
Specifications
Specifications
2 mL
50-100 nmol/ml settled resin, or up to 3 mg of 30 kDa protein per ml settled resin