Learn More
MilliporeSigma™ Novagen™ Strep·Tactin™ Superflow™ Agarose
Used for gravity flow, low pressure and FPLC chromatography.
$254.00 - $967.50
Specifications
Binding Capacity 50-100 nmol/ml settled resin, or up to 3 mg of 30 kDa protein per ml settled resin
View More Specs
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
71-592-3
View Documents
MilliporeSigma™
715923
2 mL
Each for $254.00
Add to cart
71-592-4
View Documents
MilliporeSigma™
715924
10 mL
Each for $967.50
Add to cart
Description
Description
  • Cross-linked agarose derivatized with StrepTactin protein
  • Binding capacity: 50 to 100nmol/mL settled resin (3mg of 30 kd protein per mL settled resin)
Applications:
  • Optimized for column affinity chromatography
Specifications
Specifications
50-100 nmol/ml settled resin, or up to 3 mg of 30 kDa protein per ml settled resin