Learn More
Novus Biologicals™ NovaLume Femto Chemiluminescent Substrate (HRP), Blotting
NovaLume Femto Chemiluminescent Substrate (HRP) detects HRP on a membrane. It may be used for any blotting application utilizing horseradish peroxidase (HRP) conjugates.
Manufacturer:  Novus Biologicals™ NBP261916
Catalog No. NBP2-61916
Add to cart
Description
Description
Chemiluminescent substrate supplied as two components:
  • High signal intensity and low background
  • High sensitivity - low femtogram level detection
Specifications
Specifications
Western blotting, Immunoblotting, Dot blot
100 mL
100 mL
Store at 4°C in the dark