Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PIP™ NOVAX™ Class 00 Electrical Safety Gloves Kit
Click to view available options
Color:
Black
Orange
Size:
8
9
10
11
Description
- Includes one pair of natural rubber electrical insulating gloves (150-00-11), a pair of top grain goatskin leather glove protectors (148-1000), and an 11 in. nylon glove bag
- Class 00 insulating gloves will protect workers from up to 500 V
- Meets NFPA 70E standards for use around arc flash hazards
Specifications
Certifications/Compliance | ANSI/ASTM D120, Class 00, Type I, NFPA 70E |
Applications | Industrial |
Color | Black |
Material | Natural Rubber, Top Grain Goatskin Leather |
Product Type | Electrical Insulating Gloves Kit |
Size | 8 |
For Use With (Application) | Electrical Utility Work, Industrial Plant Maintenance, and Power Generation |
Glove Cuff Style | Straight |
Includes | Natural Rubber Electrical Insulating Gloves (150-00-11), Top Grain Goatskin Leather Glove Protectors (148-1000), and a Nylon Glove Bag (148-2136) |
Series | 150-SK-00 |
Show More |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction