Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IL-12 p40 Human ELISA Kit Non-distribution product as customer accommodation. Available on GSA/VA Contract for Federal Government customers only.

Sandwich ELISA Kit

Manufacturer:  Invitrogen™ KAC1561

Catalog No. KAC1561


Add to cart

Includes: Pre-coated plate, standard, controls, antibody-HRP, diluents, wash buffer, chromogen, stop solution, and plate covers.

Description

Description

The Human Interleukin-12 p40(Hu IL-12 p40) ELISA quantitates Hu IL-12 p40 in human serum, plasma, buffered solution, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu IL-12 p40. Principle of the method The Human IL-12 p40 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

Recognizes natural and recombinant human IL-12p40.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Order Info

Shipping Condition: Wet ice

Specifications

Specifications

P29460
1.5 pg/mL
Colorimetric
3593
6.7%
Pre-coated 96 well plate,Standard,Sample diluent,Incubation Buffer,HRP conjugated Detection Antibody,Conjugate Diluent,Wash Buffer,Chromogen,Stop Solution,Controls
1.5pg/mL
96 Tests
Plasma,Serum,Supernatant
2°C to 8°C
Human
4 hr. 30 min.
15-1300 pg/mL
HRP
Colorimetric Microplate Reader
CLMF,CLMF2,IL-12B,IMD28,IMD29,NKSF,NKSF2
4%
HRP
ELISA Kit
RUO
Plasma, 100 μL|Serum, 100 μL|Supernatant, 100 μL
Human
1 hr. 20 min.
SDS
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only. Not for use in diagnostic procedures.