Learn More
Invitrogen™ Novex™ Reversible Membrane Protein Stain Kit Non-distribution product as customer accommodation. Available on GSA/VA Contract for Federal Government customers only.
Used for determining protein transfer efficiency after blotting
Manufacturer:  Invitrogen™ IB7710
Catalog No. IB7710
Add to cart
Includes: Stain Solution: 176mL; Destain Solution 150mL; Eraser Solution: 150mL.
Description
Description
Quantitative and completely reversible, this stain is compatible for use with both nitrocellulose (NC) and polyvinylidene difluoride (PVDF) membranes.
Protein Gel Electrophoresis, Protein Gel Staining and Imaging, Proteins, Expression, Isolation and Analysis, Western Blotting
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Order Info
Shipping Condition: Room Temperature
Specifications
Specifications
Stain Solution: 176 ml; Destain Solution 150 ml; Eraser Solution: 150 ml. Store at room temperature
Novex® Reversible Membrane Protein Stain
Room Temperature
Colorimetric
In-Blot Detection
Novex™
Protein
Documents
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only. Not for use in diagnostic procedures.