Learn More
Novus Biologicals™ Antibody Concentration and Clean Up Antibody Purification Kit
Allows quick and easy concentration of antibodies and proteins
$397.50
Specifications
Product Type Specs Antibody Concentration and Clean Up Antibody Purification Kit
View More Specs
Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
86-100-10
View Documents
Novus Biologicals™
8610010
3 Tests
Each for $397.50
Add to cart
Description
Description
The antibody clean up kit method utilizes a simple spin column to easily and quickly remove excess buffer from the antibody thereby providing a more concentrated antibody solution. The clean up kit also allows the experimenter to perform a simple buffer exchange to transfer the antibody into a more favorable conjugation buffer. The kit can also be used to reduce the concentration of many unwanted additives often found in antibody formulations such as azide, glycine or tris.
Specifications
Specifications
Antibody Concentration and Clean Up Antibody Purification Kit