Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ ACADM Protein
Highly purified. Generating reliable and reproducible results.
Supplier: Novus Biologicals™ NBP1452680.25MG
Description
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 26-421 of Human ACADM The Recombinant Human ACADM Protein is derived from E. coli. The Recombinant Human ACADM Protein has been validated for the following applications: SDS-Page.Specifications
0.5mg/mL | |
20mM Tris-HCl buffer (pH 8.0) containing 20% glycerol 0.1M NaCl | |
45.9kDa | |
0.25mg | |
MGSSHHHHHH SSGLVPRGSH MKANRQREPG LGFSFEFTEQ QKEFQATARK FAREEIIPVA AEYDKTGEYP VPLIRRAWEL GLMNTHIPEN CGGLGLGTFD ACLISEELAY GCTGVQTAIE GNSLGQMPII IAGNDQQKKK YLGRMTEEPL MCAYCVTEPG AGSDVAGIKT KAEKKGDEYI INGQKMWITN GGKANWYFLL ARSDPDPKAP ANKAFTGFIV EADTPGIQIG RKELNMGQRC SDTRGIVFED VKVPKENVLI GDGAGFKVAM GAFDKTRPVV AAGAVGLAQR ALDEATKYAL ERKTFGKLLV EHQAISFMLA EMAMKVELAR MSYQRAAWEV DSGRRNTYYA SIAKAFAGDI ANQLATDAVQ ILGGNGFNTE YPVEKLMRDA KIYQIYEGTS QIQRLIVARE HIDKYKN | |
RUO | |
Unconjugated |
ELISA, SDS-PAGE | |
34 | |
Protein | |
Human | |
Store at -80°C. Avoid freeze-thaw cycles. | |
ACADM | |
>90% |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction