Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Adenosylhomocysteinase/AHCY Protein

Highly purified. Generating reliable and reproducible results.

Manufacturer:  Novus Biologicals™ NBP1452710.5MG

 View more versions of this product

Catalog No. NBP1452715


Add to cart

Description

Description

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-432 of Human Adenosylhomocysteinase/AHCY The Recombinant Human Adenosylhomocysteinase/AHCY Protein is derived from E. coli. The Recombinant Human Adenosylhomocysteinase/AHCY Protein has been validated for the following applications: SDS-Page.
Specifications

Specifications

NP_000678
ELISA, SDS-Page
191
Protein
0.5mg
AHCY, 1-432aa, Human, His tag, E. coli (NP_000678).
RUO
Unlabeled
1.0mg/mL
20mM Tris-HCl buffer (pH 8.0) containing 10% glycerol
48kDa
>95%
Recombinant human AHCY protein, fused to His-tag at N-terminus, expressed in E. coli
Store at -80°C. Avoid freeze-thaw cycles.
Adenosylhomocysteinase/AHCY
Yes
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit