Learn More
Novus Biologicals™ AGL Overexpression Lysate (Native)
Manufacturer:  Novus Biologicals™ NBP209002
Catalog No. NBP209002
Add to cart
Description
Description
The AGL Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.
Specifications
Specifications
The lysate was created in HEK293T cells, using plasmid ID RC215316 and based on accession number NP_634. The protein contains a C-terminal DDK tag.
RIPA buffer
Homo sapiens AGL, mRNA.
Native
0.1mg
Protein
-80°C. Avoid freeze-thaw cycles.
Western Blot