Learn More
Novus Biologicals™ AMBRA1 Peptide
Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition
Manufacturer:  Novus Biologicals™ NBP176895PEP
Catalog No. NBP176895E
Add to cart
Description
Description
A blocking peptide containing 18 amino acids near the amino terminus of human Ambra1. Source: Synthetic (Accession #: Q9C0C7) The AMBRA1 Blocking Peptide has been validated for the following applications: Antibody Competition.
Specifications
Specifications
55626
For Blocking the binding of NBP1-76895
PBS pH 7.2 (10mM NAH2PO4, 10mM Na2HPO4, 130mM NaCl) containing 0.1% bovine serum albumin.
RUO
18 amino acids near the amino terminus of human Ambra1
0.05mg
Synthetic
Human
−20°C. Avoid freeze-thaw cycles.
AMBRA1