Learn More
Novus Biologicals™ Annexin A4 Protein
Highly purified. Generating reliable and reproducible results.
Manufacturer:  Novus Biologicals™ NBP137082
Catalog No. NBP137082
Add to cart
Description
Description
An un-tagged recombinant protein corresponding to the amino acids 1-321 of Human Annexin A4 The Recombinant Human Annexin A4 Protein is derived from E. coli. The Recombinant Human Annexin A4 Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
NP_001144
20mM Tris-HCl buffer (pH8.0), 20% glycerol, 0.2 M NaCl
36kDa
Protein
MAMATKGGTV KAASGFNAME DAQTLRKAMK GLGTDEDAII SVLAYRNTAQ RQEIRTAYKS TIGRDLIDDL KSELSGNFEQ VIVGMMTPTV LYDVQELRRA MKGAGTDEGC LIEILASRTP EEIRRISQTY QQQYGRSLED DIRSDTSFMF QRVLVSLSAG GRDEGNYLDD ALVRQDAQDL YEAGEKKWGT DEVKFLTVLC SRNRNHLLHV FDEYKRISQK DIEQSIKSET SGSFEDALLA IVKCMRNKSA YFAEKLYKSM KGLGTDDNTL IRVMVSRAEI DMLDIRAHFK RLYGKSLYSF IKGDTSGDYR KVLLVLCGGD D
ELISA, SDS-Page
307
Annexin A4 Protein
>90%
−80°C. Avoid freeze-thaw cycles.