Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ ARG2 Protein

Highly purified. Generating reliable and reproducible results.

Manufacturer:  Novus Biologicals™ NBP1452560.05MG

 View more versions of this product

Catalog No. NBP14525605


Add to cart

Description

Description

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 23-354 of Human ARG2 The Recombinant Human ARG2 Protein is derived from E. coli. The Recombinant Human ARG2 Protein has been validated for the following applications: SDS-Page.
Specifications

Specifications

0.5mg/mL
In 20mM Tris-HCl Buffer (pH 8.0) containing 10% Glycerol
38.3kDa
>95%
Human
Store at -80°C. Avoid freeze-thaw cycles.
ARG2
ELISA, SDS-Page
384
Protein
0.05mg
MGSSHHHHHH SSGLVPRGSH MVHSVAVIGA PFSQGQKRKG VEHGPAAIRE AGLMKRLSSL GCHLKDFGDL SFTPVPKDDL YNNLIVNPRS VGLANQELAE VVSRAVSDGY SCVTLGGDHS LAIGTISGHA RHCPDLCVVW VDAHADINTP LTTSSGNLHG QPVSFLLREL QDKVPQLPGF SWIKPCISSA SIVYIGLRDV DPPEHFILKN YDIQYFSMRD IDRLGIQKVM ERTFDLLIGK RQRPIHLSFD IDAFDPTLAP ATFTPVVGGL TYREGMYIAE EIHNTGLLSA LDLVEVNPQL ATSEEEAKTT ANLAVDVIAS SFGQTREGGH IVYDQLPTPS SPDESENQAR VRI
RUO
Unlabeled
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit