Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ ARL5A Protein
Highly purified. Generating reliable and reproducible results. Applications: Dot Blot, SDS-Page
$494.50 - $1472.00
Specifications
Concentration | 1mg/mL |
---|---|
For Use With (Application) | Dot Blot, SDS-PAGE |
Formulation | Liquid. In 20mM Tris-HCl buffer (pH 8.0) containing 2mM DTT, 20% glycerol, 0.2mM PMSF |
Molecular Weight (g/mol) | 23.3kDa |
Purification Method | SDS-PAGE |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP15090005
|
Novus Biologicals™
NBP1509000.05MG |
0.05mg |
Each for $494.50
|
|
|||||
NBP15090025
|
Novus Biologicals™
NBP1509000.25MG |
0.25mg |
Each for $1,472.00
|
|
|||||
Description
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-179 of Human ARL5A The Recombinant Human ARL5A Protein is derived from E. coli. The Recombinant Human ARL5A Protein has been validated for the following applications: Dot Blot, SDS-Page.Specifications
1mg/mL | |
Liquid. In 20mM Tris-HCl buffer (pH 8.0) containing 2mM DTT, 20% glycerol, 0.2mM PMSF | |
SDS-PAGE | |
Store at -80°C. Avoid freeze-thaw cycles. | |
ARL5A | |
>90% |
Dot Blot, SDS-PAGE | |
23.3kDa | |
Human | |
RUO | |
Unconjugated |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title