Learn More
Novus Biologicals™ ATG16L1 Peptide
Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition
Manufacturer:  Novus Biologicals™ NBP176863PEP
Catalog No. NBP176863E
Add to cart
Description
Description
A blocking peptide containing 18 amino acids near the center of human ATG16. Source: Synthetic (Accession #: NP_110430) The ATG16L1 Blocking Peptide has been validated for the following applications: Antibody Competition.
Specifications
Specifications
55054
For Blocking the binding of NBP1-76863
PBS pH 7.2 (10mM NAH2PO4, 10mM Na2HPO4, 130mM NaCl) containing 0.1% bovine serum albumin.
RUO
18 amino acids near the center of human ATG16
0.05mg
Synthetic
Human
−20°C. Avoid freeze-thaw cycles.
ATG16L1