Novus Biologicals™ ATPase Colorimetric Assay Kit

Colorimetric assay kit

Manufacturer: Novus BiologicalsSupplier Diversity Partner 6010122

 View more versions of this product

Catalog No. 60-101-22

  Check Availability
Add to Cart

    Specifications

    Product Type ATPase Colorimetric Assay Kit
    Quantity 5 x 480 assays
    For Use With (Application) In vitro assay
    Storage Requirements Storage is content dependent.

    Colorimetric assays for ATPases are invariably based on the formation of colored complexes between an inorganic phosphate and a dye molecule under acidic conditions. Such assays are beset with problems of reagent precipitation and high backgrounds caused by impure substrates (i.e. contaminated with inorganic phosphate) and/or by non-enzymatic (acid) hydrolysis of the substrate. Na+/K+-ATPase activity determination (PMID: 21304972)

    At the heart of our assay kits is PiColorLock Gold, a superior phosphate detection reagent which ensures high stability of the colored dye-phosphate complexes. Together with specially purified substrates and proprietary stabilizers our kits offer the lowest possible assay backgrounds and outstanding assay performance.

    Uniquely, Innova Biosciences ATPase assay kits contain highly purified ATP, which have had any free inorganic phosphate removed - this means the assays have the lowest possible background signal.

    PiColorLock Phosphate Detection Systems can also be purchased for developing simple assays for any phosphate-generating enzyme.

    This non radioactive colorimetric assay kit uses a 96 well format, and all the necessary reagents are supplied for measuring enzyme activity and are ideal for high throughput drug screening.

    Order Info

    Quantity

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit