Learn More
Novus Biologicals™ ATPase Colorimetric Assay Kit
Applications: In vitro assay
$631.35
Specifications
Product Type ATPase Colorimetric Assay Kit
View More Specs
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
60-101-20
View Documents
Novus Biologicals™
6010120
192 Tests
Each for $631.35
Add to cart
60-101-21
View Documents
Novus Biologicals™
6010121
480 Tests This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description
Description
Colorimetric assays for ATPases are invariably based on the formation of colored complexes between an inorganic phosphate and a dye molecule under acidic conditions. Such assays are beset with problems of reagent precipitation and high backgrounds caused by impure substrates (i.e. contaminated with inorganic phosphate) and/or by non-enzymatic (acid) hydrolysis of the substrate. Na+/K+-ATPase activity determination (PMID: 21304972)
At the heart of our assay kits is PiColorLock Gold, a superior phosphate detection reagent which ensures high stability of the colored dye-phosphate complexes. Together with specially purified substrates and proprietary stabilizers our kits offer the lowest possible assay backgrounds and outstanding assay performance.
Uniquely, Innova Biosciences ATPase assay kits contain highly purified ATP, which have had any free inorganic phosphate removed - this means the assays have the lowest possible background signal.
PiColorLock Phosphate Detection Systems can also be purchased for developing simple assays for any phosphate-generating enzyme.
This non radioactive colorimetric assay kit uses a 96 well format, and all the necessary reagents are supplied for measuring enzyme activity and are ideal for high throughput drug screening.
Specifications
Specifications
ATPase Colorimetric Assay Kit