Learn More
Novus Biologicals™ B7-H1/PD-L1/CD274 Peptide
Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition
Manufacturer:  Novus Biologicals™ NBP176769PEP
Catalog No. NBP176769E
Add to cart
Description
Description
A blocking peptide containing 17 amino acids near the center of human PD-L1. Source: Synthetic (Accession #: NP_054862) The PD-L1 Blocking Peptide has been validated for the following applications: Antibody Competition.
Specifications
Specifications
29126
For Blocking the binding of NBP1-76769
PBS pH 7.2 (10mM NAH2PO4, 10mM Na2HPO4, 130mM NaCl) containing 0.1% bovine serum albumin.
RUO
17 amino acids near the center of human PDL-1
0.05mg
Synthetic
Human
−20°C. Avoid freeze-thaw cycles.
B7-H1/PD-L1/CD274