Learn More
Novus Biologicals™ BACE-2 Antibody Blocking Peptide
Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition
Supplier: Novus Biologicals™ NBP177309PEP
Description
A blocking peptide containing 16 amino amino terminus of human BACE2. Source: Synthetic (Accession #: AAF17078) The BACE-2 Blocking Peptide has been validated for the following applications: Antibody Competition.Specifications
25825 | |
Store at -20 C. Avoid freeze-thaw cycles. | |
This Peptide is Useful as a Blocking Peptide for NBP1-77309. For Further Blocking Peptide Related Protocol, Click Here . | |
BACE2 | |
0.05 mg | |
Synthetic |
Human | |
PBS pH 7.2 (10 mM NaH2PO4, 10 mM Na2HPO4, 130 mM NaCl) containing 0.1% bovine serum albumin. | |
AEPLC, Asp 1, ASP1, ASP21, Aspartic-like protease 56 kDa, Aspartyl protease 1, beta secretase 2, beta-secretase 2, beta-site amyloid beta A4 precursor protein-cleaving enzyme 2, Beta-site amyloid precursor protein cleaving enzyme 2, Beta-site APP cleaving enzyme 2, beta-site APP-cleaving enzyme 2, beta-site APP-cleaving enzyme 2, EC 3.4.2310 ALP5656 kDa aspartic-like protease, CEAP1, Down region aspartic protease, Down syndrome region aspartic protease, DRAPBAE2, EC 3.4.23, EC 3.4.23.45, Memapsin-1, Membrane-associated aspartic protease 1, Theta-secretase, transmembrane aspartic proteinase Asp1 | |
BACE-2 | |
RUO - research use only | |
16 amino amino terminus of human BACE2 |