Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ BLMH/Bleomycin Hydrolase Protein

Highly purified and high bioactivity. Generating reliable and reproducible results. Applications: Functional, SDS-Page, Bioactivity

Specifications

Common Name BLMH/Bleomycin Hydrolase
Conjugate Unlabeled
For Use With (Application) Western Blot, ELISA
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP1494525
View Documents
Novus Biologicals™
NBP1494520.5MG
0.5mg This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
NBP14945201
View Documents
Novus Biologicals™
NBP1494520.1MG
0.1mg This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

A bioactive recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-455 of Human BLMH/Bleomycin Hydrolase The Recombinant Human BLMH/Bleomycin Hydrolase Protein is derived from E. coli. The Recombinant Human BLMH/Bleomycin Hydrolase Protein has been validated for the following applications: Functional, SDS-Page, Bioactivity.
Specifications

Specifications

BLMH/Bleomycin Hydrolase
Western Blot, ELISA
RUO
Unlabeled
Store at -80°C. Avoid freeze-thaw cycles.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit