Learn More
Novus Biologicals™ BMI-1 Peptide
Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition
Manufacturer:  Novus Biologicals™ NBP176685PEP
Catalog No. NBP176685E
Add to cart
Description
Description
A blocking peptide containing 15 amino acids near the center of human BMI-1. Source: Synthetic (Accession #: P35226) The BMI-1 Blocking Peptide has been validated for the following applications: Antibody Competition.
Specifications
Specifications
648
For Blocking the binding of NBP1-76685
PBS pH 7.2 (10mM NAH2PO4, 10mM Na2HPO4, 130mM NaCl) containing 0.1% bovine serum albumin.
RUO
15 amino acids near the center of human BMI-1
0.05mg
Synthetic
Human
−20°C. Avoid freeze-thaw cycles.
BMI-1