Learn More
Novus Biologicals™ BMP-5 Protein
Highly purified. Generating reliable and reproducible results.
Manufacturer:  Novus Biologicals™ NBP130182
Catalog No. NBP130182
Add to cart
Description
Description
An un-tagged recombinant protein corresponding to the amino acids 317-454 of Human BMP-5 The Recombinant Human BMP-5 Protein is derived from E. coli. The Recombinant Human BMP-5 Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
NP_066551
10mM Sodium Citrate buffer (pH 3.5), 10% glycerol
15.7kDa
Protein
MAANKRKNQN RNKSSSHQDS SRMSSVGDYN TSEQKQACKK HELYVSFRDL GWQDWIIAPE GYAAFYCDGE CSFPLNAHMN ATNHAIVQTL VHLMFPDHVP KPCCAPTKLN AISVLYFDDS SNVILKKYRN MVVRSCGCH
ELISA, SDS-Page
653
BMP-5 protein
>95%
−80°C. Avoid freeze-thaw cycles.