Learn More
Novus Biologicals™ BNIP3L Peptide
Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition
Manufacturer:  Novus Biologicals™ NBP177296PEP
Catalog No. NBP177296E
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
A blocking peptide containing 16 amino acids of human Bnip3L . Source: Synthetic (Accession #: AF067396) The BNIP3L Blocking Peptide has been validated for the following applications: Antibody Competition.
Specifications
Specifications
Human
0.05mg
Synthetic
RUO
16 amino acids of human Bnip3L
665
For blocking the activity of Bnip3L, NBP1-77296
PBS pH 7.2 (10mM NAH2PO4, 10mM Na2HPO4, 130mM NaCl) containing 0.1% bovine serum albumin.
−20°C. Avoid freeze-thaw cycles.
BNIP3L