Learn More
Novus Biologicals™ Bovine IgG, Fab Protein
Bovine Isotype Control Antibody
Manufacturer:  Novus Biologicals™ NBP197032
Catalog No. NBP197032
Add to cart
Description
Description
Bovine IgG, Fab Isotype Control antibody specifically detects Bovine IgG, Fab in Bovine samples. It is validated for Western Blot, ELISA, Immunoprecipitation.
Specifications
Specifications
WB, ELISA, IP
Bovine IgG, Fab Protein
Bovine
Bovine
0.02 M Potassium Phosphate, 0.15M Sodium Chloride pH 7.2.
4°C short term, −20°C long term. Avoid freeze-thaw cycles.
Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Bovine Serum, anti-Bovine IgG and anti-Bovine IgG F(ab)2. No reaction was observed against anti-Bovine IgG F(c) or anti- Papain.