Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ BRE Peptide

Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition

Manufacturer:  Novus Biologicals™ NBP176830PEP

Catalog No. NBP176830E


Add to cart

Description

Description

A blocking peptide containing 18 amino acids near the amino terminus of human BRCC45. Source: Synthetic (Accession #: NP_004890) The BRE Blocking Peptide has been validated for the following applications: Antibody Competition.
Specifications

Specifications

9577
For Blocking the binding of NBP1-76830
PBS pH 7.2 (10mM NAH2PO4, 10mM Na2HPO4, 130mM NaCl) containing 0.1% bovine serum albumin.
RUO
18 amino acids near the amino terminus of human BRCC45
BRE
0.05mg
Synthetic
Human
−20°C. Avoid freeze-thaw cycles.
BABAM2
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit