Learn More
Novus Biologicals™ Canine Canine IgG, Fab Protein
Canine Isotype Control Antibody
Manufacturer:  Novus Biologicals™ NBP197024
Catalog No. NBP197024
Add to cart
Description
Description
Canine IgG, Fab Isotype Control antibody specifically detects Canine IgG, Fab in Canine samples. It is validated for Western Blot, ELISA, Immunoprecipitation, SDS-Page.
Specifications
Specifications
WB, ELISA, IP
Canine IgG, Fab Protein
Canine
Canine
None.
4°C short term, −20°C long term. Avoid freeze-thaw cycles.
Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Canine IgG, anti-Canine IgG F(ab)2 and anti-Canine Serum. No reaction was observed against anti-Canine IgG F(c) or anti- Papain.