Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Carbonic Anhydrase VIII/CA8 Protein

Highly purified. Generating reliable and reproducible results.

Manufacturer:  Novus Biologicals™ NBP178887100UG

Catalog No. NBP17888710


Add to cart

Description

Description

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-290 of Human Carbonic Anhydrase VIII/CA8 The Recombinant Human Carbonic Anhydrase VIII/CA8 Protein is derived from E. coli. The Recombinant Human Carbonic Anhydrase VIII/CA8 Protein has been validated for the following applications: SDS-Page.
Specifications

Specifications

1mg/mL
Liquid. 20mM Tris-HCl buffer (pH 8.0) containing 20% glycerol, 1mM DTT
SDS-PAGE
100μg
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Carbonic Anhydrase VIII/CA8
SDS-Page
35.5kDa
>90%
Human
RUO
Unlabeled
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit