Learn More
Novus Biologicals™ Caspase-7 Peptide
Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition
Manufacturer:  Novus Biologicals™ NBP177210PEP
Catalog No. NBP177210E
Add to cart
Description
Description
A blocking peptide containing 16 amino acids near the carboxy terminus of human caspase-7. Source: Synthetic (Accession #: P55210) The Caspase-7 Blocking Peptide has been validated for the following applications: Antibody Competition.
Specifications
Specifications
840
For Blocking the binding of NBP1-77210
PBS pH 7.2 (10mM NAH2PO4, 10mM Na2HPO4, 130mM NaCl) containing 0.1% bovine serum albumin.
RUO
16 amino acids near the carboxy terminus of human caspase-7
0.05mg
Synthetic
Human
−20°C. Avoid freeze-thaw cycles.
Caspase-7