Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ CCL14/HCC-1/HCC-3 Protein

Highly purified. Generating reliable and reproducible results.

Manufacturer:  Novus Biologicals™ NBP1510070.1MG

 View more versions of this product

Catalog No. NBP15100701

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 20-93 of Human CCL14/HCC-1/HCC-3 The Recombinant Human CCL14/HCC-1/HCC-3 Protein is derived from E. coli. The Recombinant Human CCL14/HCC-1/HCC-3 Protein has been validated for the following applications: SDS-Page.
Specifications

Specifications

0.5mg/mL
Liquid. In Phosphate Buffered Saline pH 7.4 containing 10% glycerol
SDS-PAGE
0.1mg
RUO
Unlabeled
SDS-Page
10.9kDa
>95%
Store at -80°C. Avoid freeze-thaw cycles.
CCL14/HCC-1/HCC-3
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit