Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Cell Surface Toll-like Receptor Staining Flow Assay Kit: TLR4
Applications: Flow Cytometry
Supplier: Novus Biologicals™ NBP226247
Specifications
Store at 4°C. Do not freeze. | |
Staining buffer 1X 3 X 60mL, Paraformaldehyde 10% 1 X 10mL | |
Cell Surface Toll-like Receptor Staining Flow Assay Kit |
1 Kit | |
Flow Cytometry |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction