Learn More
Novus Biologicals™ CRABP1 Protein
Highly purified. Generating reliable and reproducible results.
Manufacturer:  Novus Biologicals™ NBP130183
Catalog No. NBP130183
Add to cart
Description
Description
An un-tagged recombinant protein corresponding to the amino acids 1-137 of Human CRABP1 The Recombinant Human CRABP1 Protein is derived from E. coli. The Recombinant Human CRABP1 Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
NP_004369
20mM Tris-HCl buffer (pH8.0), 10% glycerol
15.5kDa
Protein
MPNFAGTWKM RSSENFDELL KALGVNAMLR KVAVAAASKP HVEIRQDGDQ FYIKTSTTVR TTEINFKVGE GFEEETVDGR KCRSLATWEN ENKIHCTQTL LEGDGPKTYW TRELANDELI LTFGADDVVC TRIYVRE
ELISA, SDS-Page
1381
CRABP1 Protein
>95%
−80°C. Avoid freeze-thaw cycles.