Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CRMP1 Protein, Novus Biologicals™

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

$293.00 - $818.00

Specifications

Common Name CRMP1
Conjugate Unlabeled
For Use With (Application) SDS-Page
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Regulatory Status RUO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP23206201
View Documents
Novus Biologicals
NBP2320620.1MG
0.1mg
Each for $293.00
Add to cart
 
NBP2320625
View Documents
Novus Biologicals
NBP2320620.5MG
0.5mg
Each for $818.00
Add to cart
 
Description

Description

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-572 of Human CRMP1 The Recombinant Human CRMP1 Protein is derived from E. coli. The Recombinant Human CRMP1 Protein has been validated for the following applications: SDS-Page.
Specifications

Specifications

CRMP1
SDS-Page
RUO
Unlabeled
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit