Learn More
Novus Biologicals™ CRTH-2/GPR44 Peptide
Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition
Manufacturer:  Novus Biologicals™ NBP176756PEP
Catalog No. NBP176756E
Add to cart
Description
Description
A blocking peptide containing 15 amino acid peptide from near the center of human CRTH2. Source: Synthetic (Accession #: NP_004769) The CRTH-2/GPR44 Blocking Peptide has been validated for the following applications: Antibody Competition.
Specifications
Specifications
11251
For Blocking the binding of NBP1-76756
PBS pH 7.2 (10mM NAH2PO4, 10mM Na2HPO4, 130mM NaCl) containing 0.1% bovine serum albumin.
RUO
15 amino acid peptide from near the center of human CRTH2
0.05mg
Synthetic
Human
−20°C. Avoid freeze-thaw cycles.
CRTH-2/GPR44