Learn More
Novus Biologicals™ Cyclin H Overexpression Lysate
Supplier: Novus Biologicals™ NBL108883
Description
The Cyclin H Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.Specifications
The lysate was created in HEK293T cells, using plasmid ID RC24982 and based on accession number NM_1239. The protein contains a C-terminal DDK tag. | |
0.1 mg | |
Protein | |
-80°C. Avoid freeze-thaw cycles. |
Western Blot | |
RIPA buffer | |
Homo sapiens cyclin H (CCNH), mRNA. | |
Native |