Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Dimethylarginine Dimethylaminohydrolase 1/DDAH1 Protein

Highly purified. Generating reliable and reproducible results.

Manufacturer:  Novus Biologicals™ NBP1494420.1MG

 View more versions of this product

Catalog No. NBP14944201


Add to cart

Description

Description

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-285 of Human Dimethylarginine Dimethylaminohydrolase 1/DDAH1 The Recombinant Human Dimethylarginine Dimethylaminohydrolase 1/DDAH1 Protein is derived from E. coli. The Recombinant Human Dimethylarginine Dimethylaminohydrolase 1/DDAH1 Protein has been validated for the following applications: SDS-Page.
Specifications

Specifications

1mg/mL
20mM Tris-HCl buffer (pH 8.0) containing 1mM DTT, 10% glycerol, 50mM NaCl
33.5kDa
>95%
Human
Store at -80°C. Avoid freeze-thaw cycles.
Dimethylarginine Dimethylaminohydrolase 1/DDAH1
ELISA, SDS-Page
23576
Protein
0.1mg
Human DDAH1, 1- 285 aa, His tagged
RUO
Unlabeled
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit