Learn More
Novus Biologicals™ Donkey IgG, Fab2 Protein
Donkey Isotype Control Antibody
Manufacturer:  Novus Biologicals™ NBP197009
Catalog No. NBP197009
Add to cart
Description
Description
Donkey IgG, Fab2 Isotype Control antibody specifically detects Donkey IgG, Fab2 in Donkey samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation.
Specifications
Specifications
WB, ELISA, IP
Donkey IgG, Fab2 Protein
Donkey
Donkey
Potassium Phosphate, 0.15M Sodium Chloride pH 7.2.
4°C short term, −20°C long term. Avoid freeze-thaw cycles.
Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Donkey IgG, anti-Donkey IgG F(ab)2 and anti-Donkey Serum. No reaction was observed against anti-Donkey IgG F(c) or anti-Pepsin.