Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Donkey Whole Serum

Polyclonal serum without presevatives

Manufacturer:  Novus Biologicals™ NBP150088

Catalog No. NBP150088


Add to cart

Description

Description

Applications:
mmunoelectrophoresis

Recommended Storage

Store at-20°C

Specifications

Specifications

No Buffer
Immunoelectrophoresis
100mL
Donkey whole serum
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit