Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ DPPIV/CD26 Protein

Highly purified and high bioactivity. Generating reliable and reproducible results. Applications: Functional, SDS-Page, Bioactivity

$421.50 - $1081.00

Specifications

Common Name DPPIV/CD26
Conjugate Unlabeled
For Use With (Application) ELISA, SDS-Page
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBC11852901
View Documents
Novus Biologicals™
NBC1185290.1MG
0.1mg
Each for $421.50
Add to cart
 
NBC1185295
View Documents
Novus Biologicals™
NBC1185290.5MG
0.5mg
Each for $1,081.00
Add to cart
 
Description

Description

A bioactive recombinant protein with a C-Terminal His-tag and corresponding to the amino acids 39-766 of Human DPPIV/CD26 The Recombinant Human DPPIV/CD26 Protein is derived from Baculovirus. The Recombinant Human DPPIV/CD26 Protein has been validated for the following applications: Functional, SDS-Page, Bioactivity.
Specifications

Specifications

DPPIV/CD26
ELISA, SDS-Page
RUO
Unlabeled
Store at -80°C. Avoid freeze-thaw cycles.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit