Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Dynein light chain 2 cytoplasmic Protein
Highly purified. Generating reliable and reproducible results.
$398.00 - $1110.00
Specifications
For Use With (Application) | SDS-PAGE |
---|---|
Formulation | Liquid. In 20mM Tris-HCl buffer (pH 8.0) containing 1mM DTT, 30% glycerol, 0.2M NaCl |
Molecular Weight (g/mol) | 12.5kDa |
Purification Method | SDS-PAGE |
Source | Human |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP1723335
|
Novus Biologicals™
NBP1723330.5MG |
0.5mg |
Each for $1,110.00
|
|
|||||
NBP17233301
|
Novus Biologicals™
NBP1723330.1MG |
0.1mg |
Each for $398.00
|
|
|||||
Description
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-89 of Human Dynein light chain 2 cytoplasmic The Recombinant Human Dynein light chain 2 cytoplasmic Protein is derived from E. coli. The Recombinant Human Dynein light chain 2 cytoplasmic Protein has been validated for the following applications: SDS-Page.Specifications
SDS-PAGE | |
12.5kDa | |
Human | |
RUO | |
Unconjugated |
Liquid. In 20mM Tris-HCl buffer (pH 8.0) containing 1mM DTT, 30% glycerol, 0.2M NaCl | |
SDS-PAGE | |
Store at -80°C. Avoid freeze-thaw cycles. | |
Dynein light chain 2 cytoplasmic | |
>90% |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title