Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ eIF5A2 Protein
SDP

Highly purified. Generating reliable and reproducible results.

Supplier:  Novus Biologicals™ NBP1452610.25MG

 View more versions of this product

Catalog No. NBP14526102


Add to Cart

Description

Description

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-153 of Human eIF5A2 The Recombinant Human eIF5A2 Protein is derived from E. coli. The Recombinant Human eIF5A2 Protein has been validated for the following applications: SDS-Page.
Specifications

Specifications

0.5mg/mL
In 20mM Tris-HCl Buffer (pH 8.0) containing 10% Glycerol
18.9kDa
0.25mg
MGSSHHHHHH SSGLVPRGSH MADEIDFTTG DAGASSTYPM QCSALRKNGF VVLKGRPCKI VEMSTSKTGK HGHAKVHLVG IDIFTGKKYE DICPSTHNMD VPNIKRNDYQ LICIQDGYLS LLTETGEVRE DLKLPEGELG KEIEGKYNAG EDVQVSVMCA MSEEYAVAIK PCK
RUO
Unconjugated
ELISA, SDS-PAGE
56648
Protein
Human
Store at -80°C. Avoid freeze-thaw cycles.
eIF5A2
>85%
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit