Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ EMG1 Protein
SDP

Highly purified. Generating reliable and reproducible results.

$398.00 - $1110.00

Specifications

Concentration 1mg/mL
For Use With (Application) SDS-PAGE
Formulation Liquid. In 20mM Tris-HCl buffer (pH 8.0) containing 0.1M NaCl, 10% glycerol, 1mM DTT
Purification Method SDS-PAGE
Source Human
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP1989145
SDP
View Documents
Novus Biologicals™
NBP1989140.5MG
0.5mg
Each for $1,110.00
Add to Cart
 
NBP19891401
SDP
View Documents
Novus Biologicals™
NBP1989140.1MG
0.1mg
Each for $398.00
Add to Cart
 
Description

Description

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-244 of Human EMG1 The Recombinant Human EMG1 Protein is derived from E. coli. The Recombinant Human EMG1 Protein has been validated for the following applications: SDS-Page.
Specifications

Specifications

1mg/mL
Liquid. In 20mM Tris-HCl buffer (pH 8.0) containing 0.1M NaCl, 10% glycerol, 1mM DTT
Human
RUO
Unconjugated
SDS-PAGE
SDS-PAGE
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
EMG1
>95%
SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit