Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ ERP44 Protein
Highly purified. Generating reliable and reproducible results.
Supplier: Novus Biologicals™ NBP1483240.1MG
Description
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 30-406 of Human ERP44 The Recombinant Human ERP44 Protein is derived from E. coli. The Recombinant Human ERP44 Protein has been validated for the following applications: SDS-Page.Specifications
1mg/mL | |
Liquid. In 20mM Tris-HCl (pH 7.5) containing 10% Glycerol | |
Protein | |
Human | |
Store at -80°C. Avoid freeze-thaw cycles. | |
ERP44 | |
>90% |
ELISA, SDS-PAGE | |
48.0kDa | |
0.1mg | |
Human ERP44, 30-406a.a. Sequence: MRGSHHHHHHGMASMTGGQQMGRDLYDDDDKDRWAGSMEITSLDTENIDEILNNADVALVNFYADWCRFSQMLHPIFEEASDVIKEEFPNENQVVFARVDCDQHSDIAQRYRISKYPTLKLFRNGMMMKR EYRGQRSVKALADYIRQQKSDPIQEIRDLAEITTLDRSKRNIIGYFEQKDSDNYRVFERVANILHDDCAFLSAFGDVSKPERYSGDNII | |
RUO | |
Unconjugated |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction