Learn More
Novus Biologicals™ FABP7/B-FABP Peptide
Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition
Manufacturer:  Novus Biologicals™ NBP176804PEP
Catalog No. NBP176804E
Add to cart
Description
Description
A blocking peptide containing 17 amino acids near the center of human FABP7. Source: Synthetic (Accession #: EAW48166) The FABP7/B-FABP Blocking Peptide has been validated for the following applications: Antibody Competition.
Specifications
Specifications
2173
For Blocking the binding of NBP1-76804
PBS pH 7.2 (10mM NAH2PO4, 10mM Na2HPO4, 130mM NaCl) containing 0.1% bovine serum albumin.
RUO
17 amino acids near the center of human FABP7
0.05mg
Synthetic
Human
−20°C. Avoid freeze-thaw cycles.
FABP7/B-FABP