Learn More
Novus Biologicals™ FAIM2 Peptide
Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition
Manufacturer:  Novus Biologicals™ NBP177298PEP
Catalog No. NBP177298E
Add to cart
Description
Description
A blocking peptide containing 16 amino acids near the amino terminus of human FAIM2. Source: Synthetic (Accession #: AAF06327) The FAIM2 Blocking Peptide has been validated for the following applications: Antibody Competition.
Specifications
Specifications
23017
For Blocking the binding of NBP1-77298
PBS pH 7.2 (10mM NAH2PO4, 10mM Na2HPO4, 130mM NaCl) containing 0.1% bovine serum albumin.
RUO
16 amino acids near the amino terminus of human LFG
0.05mg
Synthetic
Human
−20°C. Avoid freeze-thaw cycles.
FAIM2