Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Fibrillarin Protein

Manufacturer:  Novus Biologicals™ NBP1485940.05MG

 View more versions of this product

Catalog No. NBP14859405

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

0.5mg/mL
Liquid. In 20mM Tris-HCl buffer (pH 8.0) containing 20% glycerol, 1mM DTT, 0.2M NaCl, 1mM EDTA.
28.9kDa
>85%
Human
Store at -80°C. Avoid freeze-thaw cycles.
Fibrillarin
ELISA, SDS-Page
2091
Protein
0.05mg
FBL, 83-321 aa. sequence: MGSSHHHHHHSSGLVPRGSHRSMGKNVMVEPHRHEGVFICRGKEDALVTKNLVPGESVYGEKRVSISEGDDKIEYRAWNPFRSKLAAAILGGVDQIHIKPGAKVLYLGAASGTTVSHVSDIVGPDGLVYAVEFSHRSGRDLINLAKKRTNIIPVIEDARHPHKYRMLIAMVDVIFADVAQPDQTRIVALNAHTFLRNGGHFVISIKANCIDSTASAEAVFASEVKKMQQENMKPQEQLTLEPYERDHAVVVGVYRPPPKVKN
RUO
Unlabeled
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit